Anti-KIF20A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-7750R-CY5.5
10468-866EA 1270.19 CAD
10468-866
Anti-KIF20A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIF20A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR