Anti-PRMT5 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-6992R-CY5
10459-268EA 1270.19 CAD
10459-268
Anti-PRMT5 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. Specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins Sm D1 (SNRPD1) and Sm D3 (SNRPD3); such methylation being required for the assembly and biogenesis of snRNP core particles. Methylates SUPT5H. Mono- and dimethylates arginine residues of myelin basic protein (MBP) in vitro. Plays a role in the assembly of snRNP core particles. May play a role inMore Product Information