Anti-FASTKD2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-6775R-FITC
10454-454EA 1270.19 CAD
10454-454
Anti-FASTKD2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FASTKD2 belongs to the FAST kinase family and it contains 1 RAP domain. There are two isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FASTKD2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol FASTKD2
Environmentally Preferable
Antigen name FAST kinase domains 2
Antigen synonyms KIAA0971
Antibody type Primary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 22868
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR