Anti-A1CF Rabbit Polyclonal Antibody

Supplier: Bioss

BS-6773R
10454-402EA 997.44 CAD
10454-402
Anti-A1CF Rabbit Polyclonal Antibody
Antibodies
Essential component of the apolipoprotein B mRNA editing enzyme complex which is responsible for the postranscriptional editing of a CAA codon for Gln to a UAA codon for stop in APOB mRNA. Binds to APOB mRNA and is probably responsible for docking the catalytic subunit, APOBEC1, to the mRNA to allow it to deaminate its target cytosine. The complex also protects the edited APOB mRNA from nonsense-mediated decay.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: A1CF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR