Anti-KCNQ1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-6760R-CY5
10454-158EA 1270.21 CAD
10454-158
Anti-KCNQ1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Probably important in cardiac repolarization. Associates with KCNE1 (MinK) to form the I(Ks) cardiac potassium current. Elicits a rapidly activating, potassium-selective outward current. Muscarinic agonist oxotremorine-M strongly suppresses KCNQ1/KCNE1 current in CHO cells in which cloned KCNQ1/KCNE1 channels were coexpressed with M1 muscarinic receptors. May associate also with KCNE3 (MiRP2) to form the potassium channel that is important for cyclic AMP-stimulated intestinal secretion of chloride ions, which is reduced in cystic fibrosis and pathologically stimulated in cholera and otherMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KCNQ1
Antigen name Potassium Voltage-gated Channel KQT-like Subfamily Member 1
Environmentally Preferable
Antigen synonyms Kv7.1|JLNS1|FLJ26167|KCNQ1|potassium voltage-gated channel|Jervell and Lange-Nielsen syndrome 1|WRS|ATFB1|long (electrocardiographic) QT syndrome|SQT2|Ward-Romano syndrome 1|LQT1|LQT|KQT-like subfamily|Kv1.9|member 1|KCNA8|slow delayed rectifier channel subunit|KVLQT1|kidney and cardiac voltage dependend K+ channel|RWS|KCNA9
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Dog,Rat,Mouse
Host Rabbit
Gene ID 3784
Isotype IgG
ImmunoChemistry Yes
Immunogen 500-550/676
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR