Anti-THRSP Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6738R-CY7
10453-576EA 1270.19 CAD
10453-576
Anti-THRSP Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Plays a role in the regulation of lipogenesis, especially in lactating mammary gland. Important for the biosynthesis of triglycerides with medium-length fatty acid chains. May modulate lipogenesis by interacting with MID1IP1 and preventing its interaction with ACACA (By similarity). May function as transcriptional coactivator. May modulate the transcription factor activity of THRB.Mainly expressed in tissues that synthesize triglycerides.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: THRSP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol THRSP
Antigen name Thyroid Hormone Responsive
Environmentally Preferable
Antigen synonyms SPOT14|S14
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 7069
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR