Anti-EMD Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-6660R-CY5
10451-990EA 1270.19 CAD
10451-990
Anti-EMD Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Stabilizes and promotes the formation of a nuclear actin cortical network. Stimulates actin polymerization in vitro by binding and stabilizing the pointed end of growing filaments. Inhibits beta-catenin activity by preventing its accumulation in the nucleus. Acts by influencing the nuclear accumulation of beta-catenin through a CRM1-dependent export pathway. Links centrosomes to the nuclear envelope via a microtubule association. EMD and BAF are cooperative cofactors of HIV-1 infection. Association of EMD with the viral DNA requires the presence of BAF and viral integrase. The association ofMore Product Information