Anti-SLC3A2 Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-6659R-CY5.5
10451-968EA 1270.21 CAD
10451-968
Anti-SLC3A2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Required for the function of light chain amino-acid transporters. Involved in sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan. Involved in guiding and targeting of LAT1 and LAT2 to the plasma membrane. When associated with SLC7A6 or SLC7A7 acts as an arginine/glutamine exchanger, following an antiport mechanism for amino acid transport, influencing arginine release in exchange for extracellular amino acids. Plays a role in nitric oxide synthesis in human umbilical vein endothelial cells (HUVECs) viaMore Product Information