Anti-MMTN Rabbit Polyclonal Antibody

Supplier: Bioss

BS-6551R
10449-592EA 997.44 CAD
10449-592
Anti-MMTN Rabbit Polyclonal Antibody
Antibodies
NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene cause progressive encephalopathy resulting from mitochondrial complex I deficiency.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR