Anti-EZH1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-6528R
10448-906EA 997.44 CAD
10448-906
Anti-EZH1 Rabbit Polyclonal Antibody
Antibodies
Polycomb group (PcG) protein. Catalytic subunit of the PRC2/EED-EZH1 complex, which methylates 'Lys-27' of histone H3, leading to transcriptional repression of the affected target gene. Able to mono-, di- and trimethylate 'Lys-27' of histone H3 to form H3K27me1, H3K27me2 and H3K27me3, respectively. Required for embryonic stem cell derivation and self-renewal, suggesting that it is involved in safeguarding embryonic stem cell identity. Compared to EZH1-containing complexes, it is less abundant in embryonic stem cells, has weak methyltransferase activity and plays a less critical role inMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol EZH1
Antigen name Enhancer of zeste homolog 1
Environmentally Preferable
Antigen synonyms KMT6B|Histone-lysine N-methyltransferase EZH1|ENX-2|KIAA0388|EZH1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 2145
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR