Anti-ADI1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-6507R
10448-640EA 997.44 CAD
10448-640
Anti-ADI1 Rabbit Polyclonal Antibody
Antibodies
his gene encodes an enzyme that belongs to theaci-reductone dioxygenase family of metal-binding enzymes, whichare involved in methionine salvage. This enzyme may regulate mRNAprocessing in the nucleus, and may carry out different functionsdepending on its localization. Related pseudogenes have beendefined on chromosomes 8 and 20.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol ADI1
Environmentally Preferable
Antigen name Acireductone Dioxygenase 1
Antigen synonyms MTCBP1|Ni-ARD|ARD|APL1|HMFT1638|Fe-ARD|mtnD|SIPL
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 55256
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR