Anti-C14ORF166 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6514R-CY5.5
10448-592EA 1270.19 CAD
10448-592
Anti-C14ORF166 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
CGI-99 is a 244 amino acid protein that localizes to the nucleus, as well as to the cytoplasm, and belongs to the UPF0568 family. Expressed in a variety of tissues with particularly high levels in heart and skeletal muscle, CGI-99 functions as a homodimer that interacts with Ninein and is thought to regulate Ninein function. CGI-99 is overexpressed in brain tumor tissue, suggesting a role in tumor development and metastasis. The gene encoding CGI-99 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (More Product Information
Order Now

SPECIFICATIONS

Antigen symbol C14ORF166
Antigen name Chromosome 14 open reading frame 166
Environmentally Preferable
Antigen synonyms CGI-99|CLE7|CLE|LCRP369|CGI99|RLLM1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 51637
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR