Anti-ALKBH8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-6510R-FITC
10447-908EA 1270.19 CAD
10447-908
Anti-ALKBH8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Catalyzes the methylation of 5-carboxymethyl uridine to 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in tRNA. Catalyzes the last step in the formation of 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in target tRNA. Has a preference for tRNA(Arg) and tRNA(Glu), and does not bind tRNA(Lys). Required for normal survival after DNA damage. May inhibit apoptosis and promote cell survival and angiogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALKBH8
Clonality: Polyclonal
Clone:
Conjugation: FITC (FluoresceinMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR