Anti-CAMP Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-6433R-CY3
10446-992EA 1270.19 CAD
10446-992
Anti-CAMP Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cathelicidins are a family of antimicrobial proteins found in the peroxidase-negative granules of neutrophils. Along with the family of proteins known as defensins, cathelicidins participate in the first line of defense by preventing local infection and systemic invasion of microbes. FALL-39 precursor (FALL-39 peptide antibiotic, cationic anti-microbial protein, CAMP, CAP-18, HSD26) is a cathelicidin anti-microbial protein that contains the antibacterial peptide LL-37 (amino acids 134-170). In contrast to the defensins, which are cysteine-rich peptides that fold in ∫-pleated sheets, LL-37More Product Information