Anti-RORB Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-6216R-CY7
10433-032EA 1270.19 CAD
10433-032
Anti-RORB Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Nuclear receptor that binds DNA as a monomer to ROR response elements (RORE) containing a single core motif half-site 5'-AGGTCA-3' preceded by a short A-T-rich sequence. Considered to have intrinsic transcriptional activity, have some natural ligands such as all-trans retinoic acid (ATRA) and other retinoids which act as inverse agonists repressing the transcriptional activity. Required for normal postnatal development of rod and cone photoreceptor cells. Modulates rod photoreceptors differentiation at least by inducing the transcription factor NRL-mediated pathway. In cone photoreceptorMore Product Information