Anti-NET1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-6167R
10431-968EA 997.44 CAD
10431-968
Anti-NET1 Rabbit Polyclonal Antibody
Antibodies
This gene is part of the family of Rho guanine nucleotide exchange factors. Members of this family activate Rho proteins by catalyzing the exchange of GDP for GTP. The protein encoded by this gene interacts with RhoA within the cell nucleus and may play a role in repairing DNA damage after ionizing radiation. Pseudogenes of this gene are located on the long arms of chromosomes 1, 7 and 18. Alternative splicing results in multiple transcript variants that encode different protein isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Order Now

SPECIFICATIONS

Antigen symbol NET1
Antigen name Neuroepithelial Cell Transforming 1
Environmentally Preferable
Antigen synonyms ARHGEF8|NET1A|neuroepithelial cell transforming gene 1|NET1|Rho guanine nucleotide exchange factor (GEF) 8|p65 Net1 proto-oncogene protein|guanine nucleotide regulatory protein (oncogene)|neuroepithelioma transforming gene 1|small GTP-binding protein regulator
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 10276
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR