Anti-TP53I3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-6143R-CY7
10431-498EA 1270.19 CAD
10431-498
Anti-TP53I3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
May be involved in the generation of reactive oxygen species (ROS). Has low NADPH-dependent beta-naphthoquinone reductase activity, with a preference for 1,2-beta-naphthoquinone over 1,4-beta-naphthoquinone. Has low NADPH-dependent diamine reductase activity (in vitro).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP53I3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TP53I3
Environmentally Preferable
Antigen name Tumor protein p53 inducible protein 3
Antigen synonyms quinone oxidoreductase PIG3|quinone oxidoreductase homolog|PIG3|p53-induced gene 3 protein|tumor protein p53-inducible protein 3
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human
Host Rabbit
Gene ID 9540
Isotype IgG
ImmunoChemistry Yes
Immunogen 180-230/332
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR