Anti-ZNF431 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6120R-CY5.5
10431-064EA 1270.19 CAD
10431-064
Anti-ZNF431 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
ZNF431 belongs to the krueppel C2H2-type zinc-finger protein family and is probably involved in transcriptional regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF431
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ZNF431
Environmentally Preferable
Antigen name Zinc Finger Protein 431
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human
Host Rabbit
Gene ID 170959
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR