Anti-Rho GTPase activating protein 29 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-6098R-CY3
10430-504EA 1270.19 CAD
10430-504
Anti-Rho GTPase activating protein 29 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state. Has strong activity toward RHOA, and weaker activity toward RAC1 and CDC42. May act as a specific effector of RAP2A to regulate Rho. In concert with RASIP1, suppresses RhoA signaling and dampens ROCK and MYH9 activities in endothelial cells and plays an essential role in blood vessel tubulogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Rho GTPase activating protein 29
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR