Anti-RASAL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-6088R-FITC
10430-280EA 1270.21 CAD
10430-280
Anti-RASAL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Probable inhibitory regulator of the Ras-cyclic AMP pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RASAL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol RASAL
Antigen name RAS Protein Activator Like 1
Environmentally Preferable
Antigen synonyms RAS protein activator like 1 (GAP1 like)|GAP1 like protein|RASAL|RAS-GTPase-activating protein-like|RAS protein activator like 1|RASAL1
Antibody type Primary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 8437
Isotype IgG
ImmunoChemistry Yes
Immunogen 460-510/805
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR