Anti-FGFRL1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6049R-CY5.5
10429-360EA 1270.19 CAD
10429-360
Anti-FGFRL1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
FGFRL1 is a member of the fibroblast growth factor receptor (FGFR) family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstreamMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol FGFRL1
Environmentally Preferable
Antigen name fibroblast growth factor receptor-like 1
Antigen synonyms FGFR5|FHFR
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 53834
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR