Anti-HELLS Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-6014R-CY5
10428-546EA 1270.19 CAD
10428-546
Anti-HELLS Rabbit Polyclonal Antibody (Cy5®)
Antibodies
SMARCA6 gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HELLS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR