Anti-ABCC10 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5761R-CY3
10422-466EA 1300.77 CAD
10422-466
Anti-ABCC10 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi ATP-dependent transporter probably involved in cellular detoxification through lipophilic anion extrusion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCC10
Clonality: Polyclonal
Clone:
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol ABCC10
Antigen name ATP-binding cassette, Subfamily C (CFTR/MRP), Member 10
Environmentally Preferable
Antigen synonyms SIMRP7|EST182763|MRP7|ATP binding cassette subfamily C member 10
Antibody type Primary
Clonality Polyclonal
Conjugation Cy3®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 89845
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR