Anti-MAD2L1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-5731R-CY5.5
10422-268EA 1300.77 CAD
10422-268
Anti-MAD2L1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Mad2L1 is required for the execution of the mitotic checkpoint which monitors the process of kinetochore spindle attachment and delays the onset of anaphase when this process is not complete. It inhibits the activity of the anaphase promoting complex by sequestering CDC20 until all chromosomes are aligned at the metaphase plate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR