Anti-PTPN7 Rabbit Polyclonal Antibody
Supplier: Bioss
BS-5557R
10419-018EA 1041.88 CAD
10419-018
Anti-PTPN7 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This gene is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The non-catalytic N-terminus of this PTP can interact with MAP kinases and suppress the MAP kinase activities. This PTP was shown to be involved in the regulation of T cell antigen receptor (TCR)More Product Information