Anti-PKCB Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-5566R-CY7
10417-748EA 1300.8 CAD
10417-748
Anti-PKCB Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Calcium-activated, phospholipid- and diacylglycerol (DAG)-dependent serine/threonine-protein kinase involved in various cellular processes such as regulation of the B-cell receptor (BCR) signalosome, oxidative stress-induced apoptosis, androgen receptor-dependent transcription regulation, insulin signaling and endothelial cells proliferation. Plays a key role in B-cell activation by regulating BCR-induced NF-kappa-B activation. Mediates the activation of the canonical NF-kappa-B pathway (NFKB1) by direct phosphorylation of CARD11/CARMA1 at 'Ser-559', 'Ser-644' and 'Ser-652'. PhosphorylationMore Product Information