Anti-GFAP Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-5355R-CY7
10413-946EA 1300.8 CAD
10413-946
Anti-GFAP Rabbit Polyclonal Antibody (Cy7®)
Antibodies
GFAP, a class-III intermediate filament, is a cell-specific marker that, during the development of the central nervous system, distinguishes astrocytes from other glial cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR