Anti-IDI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-5073R-FITC
10407-878EA 1300.37 CAD
10407-878
Anti-IDI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
IDI1 is a peroxisomally localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP). These are substrates for the reactions that ultimately result in the synthesis of cholesterol. There is reduction in IPP isomerase activity in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDI1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR