Anti-CYP27A1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-5049R-CY5
10407-120EA 1300.77 CAD
10407-120
Anti-CYP27A1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Catalyzes the first step in the oxidation of the side chain of sterol intermediates; the 27-hydroxylation of 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol. Has also a vitamin D3-25-hydroxylase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP27A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR