Anti-PDIA2 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-4250R-CY3
10397-064EA 1300.77 CAD
10397-064
Anti-PDIA2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The three dimensional structure of many extracellular proteins is stabilized by the formation of disulphide bonds. Studies suggest that a microsomal enzyme known as Protein Disulphide Isomerase (PDI) is involved in disulphide-bond formation and isomerization, as well as the reduction of disulphide bonds in proteins. PDI, which catalyses disulphide interchange between thiols and protein dilsulphides, has also been referred to as thiol:protein-disulphide oxidoreductase and as glutathione:insulin transhydrogenase because of its role in reduction of disulphide bonds. The highly conservedMore Product Information