Anti-ACLY Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3929R
10395-292EA 1041.92 CAD
10395-292
Anti-ACLY Rabbit Polyclonal Antibody
Antibodies
ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variantsMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ACLY
Environmentally Preferable
Antigen name ATP Citrate Lyase
Antigen synonyms ATP citrate synthase|CLATP|ATPCL|ATP-citrate synthase|ACL|citrate cleavage enzyme|ATP-citrate (pro-S-)-lyase
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 47
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR