Anti-ATG5 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4005R-CY5.5
10394-256EA 1300.77 CAD
10394-256
Anti-ATG5 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Involved in autophagic vesicle formation. Conjugation with ATG12, through a ubiquitin-like conjugating system involving ATG7 as an E1-like activating enzyme and ATG1 as an E2-like conjugating enzyme, is essential for its function. The ATG12-ATG5 conjugate acts as an E3-like enzyme which is required for lipidation of ATG8 family proteins and their association to the vesicle membranes. Involved in mitochondrial quality control after oxidative damage, and in subsequent cellular longevity. The ATG12-ATG5 conjugate also negatively regulates the innate antiviral immune response by blocking theMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ATG5
Environmentally Preferable
Antigen name Autophagy Related 5
Antigen synonyms hAPG5|APG5|APG5L|ASP|APG5-LIKE
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 9474
Isotype IgG
ImmunoChemistry Yes
Immunogen 118-158/275
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR