Anti-FBP1 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-3980R-CY7
10391-068EA 1300.77 CAD
10391-068
Anti-FBP1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate in the presence of divalent cations, acting as a rate-limiting enzyme in gluconeogenesis. Plays a role in regulating glucose sensing and insulin secretion of pancreatic beta-cells. Appears to modulate glycerol gluconeogenesis in liver. Important regulator of appetite and adiposity; increased expression of the protein in liver after nutrient excess increases circulating satiety hormones and reduces appetite-stimulating neuropeptides and thus seems to provide a feedback mechanism to limit weight gain.
RecommendedMore Product Information