Anti-NDUFS1 Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-3890R-CY5.5
10389-370EA 1300.77 CAD
10389-370
Anti-NDUFS1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity). This is the largest subunit of complex I and it is a component of the iron-sulfur (IP) fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen:More Product Information