Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-3756R-CY3
10387-386EA 1300.77 CAD
10387-386
Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Cy3®)
Antibodies
PPP1A is a serine/threonine protein phosphatase catalytic subunit that is essential for regulating cellular stress responses in eukaryotes. It binds to magnesium or manganese ions and exists as a monomer. It is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1A is involved in the regulation long term synaptic plasticity and may play an important role in dephosphorylating substrates such as Ca2+/calmodulin dependent protein kinase II.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PP1More Product Information