Anti-PKC alpha/beta/gamma Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-3531R-CY7
10374-858EA 1300.77 CAD
10374-858
Anti-PKC alpha/beta/gamma Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Protein Kinase c alpha (PKC alpha) is an 77 kDa member of the conventional group (cPKCs: sensitive to calcium, diacylglycerol, phosphatidylserine and phorbol esters) of the PKC family of serine/ threonine kinases that are involved in a wide range of physiological processes including mitogenesis, cell survival and transcriptional regulation. PKC alpha is an ubiquitously expressed PKC isozyme that has been implicated in the regulation of a broad range of cellular functions including proliferation, differentiation, development, migration, cell cell adhesion, cell extracellular matrix adhesion,More Product Information