Anti-SLC44A1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3667R
10371-582EA 1044.98 CAD
10371-582
Anti-SLC44A1 Rabbit Polyclonal Antibody
Antibodies
Choline transporter. May be involved in membrane synthesis and myelin production.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC44A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR