Anti-ALOX5 Rabbit Polyclonal Antibody
Supplier: Bioss
BS-3251R
10369-352EA 1041.85 CAD
10369-352
Anti-ALOX5 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a member of the lipoxygenase gene family and plays a dual role in the synthesis of leukotrienes from arachidonic acid. The encoded protein, which is expressed specifically in bone marrow-derived cells, catalyzes the conversion of arachidonic acid to 5(S)-hydroperoxy-6-trans-8,11,14-cis-eicosatetraenoic acid, and further to the allylic epoxide 5(S)-trans-7,9-trans-11,14-cis-eicosatetrenoic acid (leukotriene A4). Leukotrienes are important mediators of a number of inflammatory and allergic conditions. Mutations in the promoter region of this gene lead to a diminished responseMore Product Information