Anti-ALOX5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-3252R-FITC
10366-290EA 1300.8 CAD
10366-290
Anti-ALOX5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Catalyzes the first step in leukotriene biosynthesis, and thereby plays a role in inflammatory processes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALOX5
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR