Anti-ERBB4 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-3220R-CY7
10354-466EA 1300.39 CAD
10354-466
Anti-ERBB4 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This gene is a member of the Tyr protein kinase family and the epidermal growth factor receptor subfamily. It encodes a single-pass type I membrane protein with multiple cysteine rich domains, a transmembrane domain, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif. The protein binds to and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment. Mutations inMore Product Information