Anti-JUN Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3210R-CY5.5
10354-296EA 1300.39 CAD
10354-296
Anti-JUN Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. Promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol JUN
Environmentally Preferable
Antigen name c-Jun
Antigen synonyms Transcription factor AP-1|V-jun avian sarcoma virus 17 oncogene homolog|JUN|AP1|p39|Proto-oncogene c-jun|Activator protein 1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 3725
Isotype IgG
ImmunoChemistry Yes
Modification phospho-Ser63
Immunogen 50-80/331
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR