Anti-RBPJK Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-2966R-CY5
10349-178EA 1300.77 CAD
10349-178
Anti-RBPJK Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Transcriptional regulator that plays a central role in Notch signaling, a signaling pathway involved in cell-cell communication that regulates a broad spectrum of cell-fate determinations. Acts as a transcriptional repressor when it is not associated with Notch proteins. When associated with some NICD product of Notch proteins (Notch intracellular domain), it acts as a transcriptional activator that activates transcription of Notch target genes. Probably represses or activates transcription via the recruitment of chromatin remodeling complexes containing histone deacetylase or histoneMore Product Information