Anti-ITGA2B Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2636R-CY3
10343-430EA 1300.77 CAD
10343-430
Anti-ITGA2B Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Integrin alpha-IIb/beta-3 is a receptor for fibronectin, fibrinogen, plasminogen, prothrombin, thrombospondin and vitronectin. It recognizes the sequence R-G-D in a wide array of ligands. It recognizes the sequence H-H-L-G-G-G-A-K-Q-A-G-D-V in fibrinogen gamma chain. Following activation integrin alpha-IIb/beta-3 brings about platelet/platelet interaction through binding of soluble fibrinogen. This step leads to rapid platelet aggregation which physically plugs ruptured endothelial cell surface.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITGA2B
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR