Anti-IL18R1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-2615R-FITC
10342-832EA 1300.37 CAD
10342-832
Anti-IL18R1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Receptor for interleukin 18 (IL-18). Binding to the agonist leads to the activation of NF-kappa-B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL18R1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR