Anti-IL3RA Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-2600R-CY7
10342-524EA 1300.77 CAD
10342-524
Anti-IL3RA Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The protein encoded by this gene is an interleukin 3 specific subunit of a heterodimeric cytokine receptor. The receptor is comprised of a ligand specific alpha subunit and a signal transducing beta subunit shared by the receptors for interleukin 3 (IL3), colony stimulating factor 2 (CSF2/GM-CSF), and interleukin 5 (IL5). The binding of this protein to IL3 depends on the beta subunit. The beta subunit is activated by the ligand binding, and is required for the biological activities of IL3. This gene and the gene encoding the colony stimulating factor 2 receptor alpha chain (CSF2RA) form aMore Product Information