Anti-JAK2 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-2485R-CY3
10339-800EA 1300.8 CAD
10339-800
Anti-JAK2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
JAK2 (Janus Activating Kinase 2) is a tyrosine kinase of the non-receptor type, that associates with the intracellular domains of cytokine receptors; JAK2 is the predominant JAK kinase activated in response to several growth factors and cytokines such as IL-3, GM-CSF and erythropoietin; it has been found to be constitutively associated with the prolactin receptor and is required for responses to gamma interferon. Ligand binding to a variety of cell surface receptors (e.g., cytokine, growth factor, GPCRs) leads to an association of those receptors with JAK proteins, which are then activatedMore Product Information