Anti-BDKRB2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2422R-CY7
10339-490EA 1300.77 CAD
10339-490
Anti-BDKRB2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor for bradykinin. It is associated with G proteins that activate a phosphatidylinositol-calcium second messenger system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BDKRB2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol BDKRB2
Antigen name Bradykinin Receptor B2
Environmentally Preferable
Antigen synonyms BKR2|BK2|BRB2|BK-2|B2R
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 624
Isotype IgG
ImmunoChemistry Yes
Immunogen 119-131/391
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR