Anti-PPIL1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-2398R
10339-300EA 1041.92 CAD
10339-300
Anti-PPIL1 Rabbit Polyclonal Antibody
Antibodies
Peptidyl-prolyl cis-trans isomerase (PPIL1) is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Order Now

SPECIFICATIONS

Antigen symbol PPIL1
Environmentally Preferable
Antigen name Peptidylprolyl Isomerase (cyclophilin)-like 1
Antigen synonyms hCyPX|CGI-124|CYPL1|PPIase
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 51645
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR