Anti-RAP2B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2330R-CY5.5
10338-984EA 1300.77 CAD
10338-984
Anti-RAP2B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
RAP2B belongs to a family of RAS-related GTP-binding proteins. RAP proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. However, at their 61st amino acid the glutamine in RAS proteins is replaced by threonine in RAP proteins. RAP2A interacts with phospholipase C, epsilon 1 (PLCE1).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAP2B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol RAP2B
Environmentally Preferable
Antigen name Ras-related protein Rap-2b
Antigen synonyms Ras related protein RAP2B |Ras related protein RAP 2B|RAP 2B|RAP2B member of RAS oncogene family|Small GTP binding protein.RAP2B_HUMAN|Ras family small GTP binding protein RAP2B
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 5912
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR