Anti-DGAT1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2332R-CY7
10331-058EA 1300.77 CAD
10331-058
Anti-DGAT1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Catalyzes the terminal and only committed step in triacylglycerol synthesis by using diacylglycerol and fatty acyl CoA as substrates. In contrast to DGAT2 it is not essential for survival. May be involved in VLDL (very low density lipoprotein) assembly. In liver, plays a role in esterifying exogenous fatty acids to glycerol. Functions as the major acyl-CoA retinol acyltransferase (ARAT) in the skin, where it acts to maintain retinoid homeostasis and prevent retinoid toxicity leading to skin and hair disorders.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DGAT1
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol DGAT1
Environmentally Preferable
Antigen name Diacylglycerol O-acyltransferase 1
Antigen synonyms ARGP1|Acyl coenzyme A|DIAR7|Diglyceride acyltransferase|ARAT|DGAT|EC 2.3.1.20|Diacylglycerol O-acyltransferase 1|ACT-related gene product 1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 8694
Isotype IgG
ImmunoChemistry Yes
Immunogen 285-330/488
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR